Learn More
Enzo Life Sciences S5a (human) polyclonal antibody (25 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW10450025
Encompass_Preferred
Immunogen: Full-length recombinant GST-tagged human S5a (Accession number: P55036). Source: Purified from rabbit serum. Species reactivity: Human. Specificity: Recognizes the Rpn10/S5a subunit of the 19S regulator complex. Applications: WB. Purity: Purified by immunodepletion of GST-reactive antibodies followed by Protein A purification. Formulation: Liquid. In PBS containing 0.01% sodium azide.
Catalog No. 50-201-1563