Learn More
Echelon Biosciences Research Labs S6 Phosphate Acceptor Peptide - 5 mg, CAS#: 93674-74-9
SDP
Supplier:  Echelon Biosciences Research Labs 890725MG
Encompass_Preferred
S6 Phosphate Acceptor Peptide - 5mg , Arg-Arg-Leu-Ser-Ser-Leu-Arg-Ala-OH This item is non returnable
Catalog No. 50-190-9113