Learn More
Petroleum Analyzer Corporation CRM for Viscosity Testing S60 500 M
SDP
Catalog No. NC3377167
Encompass
CRM Viscosity S60, appr. 54 mm²/s @40°C; 8 mm²/s @ 100°C, 500ml
Catalog No. NC3377167 Supplier Petroleum Analyzer Corporation Supplier No. 0100044504
CRM Viscosity S60, appr. 54 mm²/s @40°C; 8 mm²/s @ 100°C, 500ml