Learn More
Electron Microscopy Sciences S60 Howard Cell for Fruit Juices
SDP
Catalog No. 5031231
Encompass_Preferred
The Howard cell is a glass slide 76 mm x 35 mm with a central circular island and is used for counting mold fibers and spores in fruit juices, especially from tomatoes. With the K20 coverglass in place a 0.1 mm thickness of liquid is contained over the central island. The coverglass has 25 calibrated fields of 1.382 mm diameter through which to view the particles. This coverglass removes the necessity of precise adjustment of the microscope and eyepiece graticule in the original Howard method.
Catalog No. 50-312-31 Supplier Electron Microscopy Sciences Supplier No. 6805160
The Howard cell is a glass slide 76 mm x 35 mm with a central circular island and is used for counting mold fibers and spores in fruit juices, especially from tomatoes. With the K20 coverglass in place a 0.1 mm thickness of liquid is contained over the central island. The coverglass has 25 calibrated fields of 1.382 mm diameter through which to view the particles. This coverglass removes the necessity of precise adjustment of the microscope and eyepiece graticule in the original Howard method.