Learn More
Petroleum Analyzer Corporation CRM for Viscosity Test S600 500 Ml
SDP
Catalog No. NC3377170
Encompass
CRM Viscosity S600, appr. 520 mm²/s @40°C; 37 mm²/s @ 100°C, 500ml
Catalog No. NC3377170 Supplier Petroleum Analyzer Corporation Supplier No. 0100044506
CRM Viscosity S600, appr. 520 mm²/s @40°C; 37 mm²/s @ 100°C, 500ml