Learn More
Beckman Coulter S6096 Swinging-Bucket Aluminum Rotor with Microplates
Catalog No. NC1618534
Encompass
S6096 Swinging-Bucket Rotor (Unshielded), Anodized Aluminum. Major applications: Serial dilution of small liquid volumes. Plates: 2 x 3 Microplates and 2 x 1 Deep-Well plates. For use in Allegra X-30 Series centrifuges.
Catalog No. NC1618534 Supplier Beckman Coulter Supplier No. B01430
S6096 Swinging-Bucket Rotor (Unshielded), Anodized Aluminum. Major applications: Serial dilution of small liquid volumes. Plates: 2 x 3 Microplates and 2 x 1 Deep-Well plates. For use in Allegra X-30 Series centrifuges.