Learn More
Medchemexpress LLC HY-100741 10mg , S63845 CAS:1799633-27-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10074110MG
Encompass_Preferred
HY-100741 10mg S63845 CAS: 1799633-27-4 S63845 is a potent and selective myeloid cell leukemia 1 (MCL1) inhibitor with a Kd of 0.19 nM for human MCL1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9017