Learn More
Medchemexpress LLC HY-100741 100mg , S63845 CAS:1799633-27-4 Purity:98%
SDP
Catalog No. 501492689
Encompass_Preferred
Medchemexpress, HY-100741 100mg S63845 CAS:1799633-27-4 Formula:C39H37ClF4N6O6S Kd: 0.19 nM (MCL1) Purity:98% S63845 is a potent and selective myeloid cell leukemia 1 ( MCL1 ) inhibitor; binds human MCL1 with a K d of 0.19 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2689 Supplier Medchemexpress LLC Supplier No. HY100741100MG
Medchemexpress, HY-100741 100mg S63845 CAS:1799633-27-4 Formula:C39H37ClF4N6O6S Kd: 0.19 nM (MCL1) Purity:98% S63845 is a potent and selective myeloid cell leukemia 1 ( MCL1 ) inhibitor; binds human MCL1 with a K d of 0.19 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.