Learn More
Selleck Chemical LLC S63845 1mg 1799633-27-4
SDP
Supplier:  Selleck Chemical LLC S83831MG
Encompass_Preferred
S63845 is a new, selective MCL-1 inhibitor with the Kd value of 0.19 nM and has no discernible binding to the other BCL-2 members, BCL-2 or BCL-XL. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1423275