Learn More
Medchemexpress LLC S63845 Solution, 10 mM * 1 mL in DMSO
SDP
Catalog No. NC2955860
Encompass_Preferred
S63845 is a potent and selective myeloid cell leukemia 1 (MCL1) inhibitor with a Kd of 0.19 nM for human MCL1.
Catalog No. NC2955860 Supplier Medchemexpress LLC Supplier No. HY10074110MMS
S63845 is a potent and selective myeloid cell leukemia 1 (MCL1) inhibitor with a Kd of 0.19 nM for human MCL1.