Learn More
Medchemexpress LLC S65487 (VOB560) | 1644600-79-2 | 99.9% | 717.26 | 50 MG
SDP
Catalog No. 5000348862
Encompass_Preferred
S65487 (VOB560) is a potent and selective BCL-2 inhibitor and a proagent of S55746. It is active against BCL-2 mutations such as G101V and D103Y, and exhibits poor affinity with MCL-1, BFL-1, and BCL-XL. This compound induces apoptosis and demonstrates anticancer activities.
  • Potent and selective BCL-2 inhibitor
  • Active against BCL-2 mutations (G101V and D103Y)
  • Induces apoptosis and inhibits cell proliferation in various hematological cancer cell lines
  • Demonstrates anticancer activities in vivo, causing complete tumor regression
Catalog No. 50-003-48862 Supplier Medchemexpress LLC Supplier No. HY13869750MG
S65487 (VOB560) is a potent and selective BCL-2 inhibitor and a proagent of S55746. It is active against BCL-2 mutations such as G101V and D103Y, and exhibits poor affinity with MCL-1, BFL-1, and BCL-XL. This compound induces apoptosis and demonstrates anticancer activities.
  • Potent and selective BCL-2 inhibitor
  • Active against BCL-2 mutations (G101V and D103Y)
  • Induces apoptosis and inhibits cell proliferation in various hematological cancer cell lines
  • Demonstrates anticancer activities in vivo, causing complete tumor regression