Learn More
Medchemexpress LLC S65487 (VOB560) | 1644600-79-2 | 99.9% | C41H41ClN6O4 | 10MG
SDP
Catalog No. 5000261641
Encompass_Preferred
S65487 (VOB560) is a potent, selective BCL-2 inhibitor used in preclinical research to study apoptosis and BCL-2-dependent cancer pathways. It shows activity against BCL-2 mutations such as G101V and D103Y and induces apoptosis in relevant cancer cell lines. The compound is supplied as a solid or as 10 mM solutions in DMSO for biochemical and cell-based assays.
  • Potent and selective BCL-2 inhibition
  • Active against BCL-2 mutations G101V and D103Y
  • Induces apoptosis and inhibits cancer cell proliferation
  • Available as solid or 10 mM solution in DMSO
  • High purity (99.9%)
  • For research use only - not for human use
Catalog No. 50-002-61641 Supplier Medchemexpress LLC Supplier No. HY13869710MG
S65487 (VOB560) is a potent, selective BCL-2 inhibitor used in preclinical research to study apoptosis and BCL-2-dependent cancer pathways. It shows activity against BCL-2 mutations such as G101V and D103Y and induces apoptosis in relevant cancer cell lines. The compound is supplied as a solid or as 10 mM solutions in DMSO for biochemical and cell-based assays.
  • Potent and selective BCL-2 inhibition
  • Active against BCL-2 mutations G101V and D103Y
  • Induces apoptosis and inhibits cancer cell proliferation
  • Available as solid or 10 mM solution in DMSO
  • High purity (99.9%)
  • For research use only - not for human use