Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC S65487 (VOB560) | 1644600-79-2 | MFCD34473226 | 99.9% | 717.26 | C41H41ClN6O4 | 5 MG
SDP

Catalog No. 5000221155
Encompass_Preferred

S65487 (VOB560) is a small-molecule BCL-2 inhibitor for research use in apoptosis and cancer biology studies. It is supplied as a high-purity solid for biochemical assays and cellular experiments, typically dissolved in DMSO. The compound is used to probe Bcl-2-mediated cell survival and resistance-associated mutations.

  • High purity (≈99.9%).
  • Molecular formula C41H41ClN6O4.
  • Molecular weight 717.26.
  • CAS number 1644600-79-2.
  • Available in small research quantities: 5 mg, 10 mg, 25 mg, 50 mg.
  • Supplied as a solid; soluble in DMSO.
  • Intended for research use only.

Catalog No. 50-002-21155 Supplier Medchemexpress LLC Supplier No. HY1386975MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S65487 (VOB560) is a small-molecule BCL-2 inhibitor for research use in apoptosis and cancer biology studies. It is supplied as a high-purity solid for biochemical assays and cellular experiments, typically dissolved in DMSO. The compound is used to probe Bcl-2-mediated cell survival and resistance-associated mutations.

  • High purity (≈99.9%).
  • Molecular formula C41H41ClN6O4.
  • Molecular weight 717.26.
  • CAS number 1644600-79-2.
  • Available in small research quantities: 5 mg, 10 mg, 25 mg, 50 mg.
  • Supplied as a solid; soluble in DMSO.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.