Learn More
Medchemexpress LLC S6K Substrate acetate | 1155.4 | 5 MG
SDP
Catalog No. 5000423237
Encompass_Preferred
S6K Substrate acetate is a substrate for active protein kinases, including CLK2, MRCKalpha, MRCKbeta, p70S6K, ROCK1, ROCK2, RSK1, RSK3, and PIM1.
  • Substrate for active protein kinases
  • Used in MAPK/ERK Pathway research
  • Suitable for various protein kinase studies
Catalog No. 50-004-23237 Supplier Medchemexpress LLC Supplier No. HYP10042A5MG
S6K Substrate acetate is a substrate for active protein kinases, including CLK2, MRCKalpha, MRCKbeta, p70S6K, ROCK1, ROCK2, RSK1, RSK3, and PIM1.
  • Substrate for active protein kinases
  • Used in MAPK/ERK Pathway research
  • Suitable for various protein kinase studies