Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC S6K Substrate acetate | 98.2% | 1155.40 (free base) | 25 MG
SDP

Catalog No. 5000426976
Encompass_Preferred

S6K Substrate acetate is a synthetic peptide substrate for active protein kinases. It is recognized by several kinases including CLK2, MRCKalpha, MRCKbeta, p70S6K, ROCK1, ROCK2, RSK1, RSK3, and PIM1.

  • Substrate for active protein kinases
  • Targeted by ribosomal S6 kinase (RSK)
  • Involves the MAPK/ERK pathway
  • Sequence: Lys-Arg-Arg-Arg-Leu-Ala-Ser-Leu-Arg (KRRRLASLR)
  • Purity of 98.2%
  • Appears as a white to off-white solid

Catalog No. 50-004-26976 Supplier Medchemexpress LLC Supplier No. 25MGHYP10042A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S6K Substrate acetate is a synthetic peptide substrate for active protein kinases. It is recognized by several kinases including CLK2, MRCKalpha, MRCKbeta, p70S6K, ROCK1, ROCK2, RSK1, RSK3, and PIM1.

  • Substrate for active protein kinases
  • Targeted by ribosomal S6 kinase (RSK)
  • Involves the MAPK/ERK pathway
  • Sequence: Lys-Arg-Arg-Arg-Leu-Ala-Ser-Leu-Arg (KRRRLASLR)
  • Purity of 98.2%
  • Appears as a white to off-white solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.