Learn More
Electron Microscopy Sciences S9 Stage Counting Grid, 0.1mm Squares
SDP
Catalog No. 5031230
Encompass_Preferred
Transmitted light stage slide with 10 x 10 grid of 0.1mm squares.
Catalog No. 50-312-30 Supplier Electron Microscopy Sciences Supplier No. 6804509
Transmitted light stage slide with 10 x 10 grid of 0.1mm squares.