Learn More
ATCC S9.6; Hybridoma; Mouse (Mus musculus)
Catalog No. 502384031
Encompass
The mAb product of this cell line may be used for microarray detection without amplification of RNA; detection of R-loops. Immunogen: DNA-RNA hybrids synthesized from φΧ174 single stranded-DNA. Primary reactivity: DNA-RNA hybrids
Catalog No. 50-238-4031 Supplier ATCC Supplier No. HB8730
The mAb product of this cell line may be used for microarray detection without amplification of RNA; detection of R-loops. Immunogen: DNA-RNA hybrids synthesized from φΧ174 single stranded-DNA. Primary reactivity: DNA-RNA hybrids