Learn More
Medchemexpress LLC S961 (acetate) | 99.83% | 4802.12 | 25 MG
SDP
Catalog No. 5000426690
Encompass_Preferred
S961 acetate is a high-affinity and selective insulin receptor (IR) antagonist. It exhibits IC50s of 0.048 nM for HIR-A, 0.027 nM for HIR-B, and 630 nM for human insulin-like growth factor I receptor (HIGF-IR) in SPA-assay. It also shows high-affinity to Rat IR and Pig IR in PEG-assay.
  • High-affinity and selective insulin receptor antagonist
  • Exhibits high affinity for human, rat, and pig IR
  • Solid, white to off-white appearance
  • Soluble in DMSO and water
  • Stable under recommended storage conditions
Catalog No. 50-004-26690 Supplier Medchemexpress LLC Supplier No. 25MGHYP2093B
S961 acetate is a high-affinity and selective insulin receptor (IR) antagonist. It exhibits IC50s of 0.048 nM for HIR-A, 0.027 nM for HIR-B, and 630 nM for human insulin-like growth factor I receptor (HIGF-IR) in SPA-assay. It also shows high-affinity to Rat IR and Pig IR in PEG-assay.
  • High-affinity and selective insulin receptor antagonist
  • Exhibits high affinity for human, rat, and pig IR
  • Solid, white to off-white appearance
  • Soluble in DMSO and water
  • Stable under recommended storage conditions