Learn More
MOLECULAR DEPOT LLC SAA A1 Recombinant - 1 mg
Catalog No. 5000013563
Encompass_Preferred
SAA Human Serum Amyloid A1 Recombinant consists of Human Serum Amyloid A1 (SAA) produced in E coli SAA is an apolipoprotein produced in the liver during cytokine-based responses to inflammation For in vitro research use only
Catalog No. 50-000-13563 Supplier MOLECULAR DEPOT LLC Supplier No. P2010014
SAA Human Serum Amyloid A1 Recombinant consists of Human Serum Amyloid A1 (SAA) produced in E coli SAA is an apolipoprotein produced in the liver during cytokine-based responses to inflammation For in vitro research use only