Learn More
Genscript Corporation SAA Antigen
Catalog No. 5000058084
Encompass
Serum Amyloid A (SAA) is an acute-phase protein The concentration of SAA in blood increases rapidly when tissue damage or inflammation occurs It is considered as an early biomarker for the diagnosis of many inflammatory diseases SAA can also be used as an important indicator of infection
Catalog No. 50-000-58084 Supplier Genscript Corporation Supplier No. T0200210
Serum Amyloid A (SAA) is an acute-phase protein The concentration of SAA in blood increases rapidly when tissue damage or inflammation occurs It is considered as an early biomarker for the diagnosis of many inflammatory diseases SAA can also be used as an important indicator of infection