Learn More
Innovative Research Inc SAA Dfcnt Hmn Pla Lyo 1mL
Catalog No. 502743988
Encompass_Preferred
Serum Amyloid A (SAA) Deficient Human Plasma Lyophilized 1mL - Serum Amyloid A (SAA) Deficient Human Plasma Lyophilized from Innovative Research is supplied as a vial of depleted human plasma in lyophilized form. The plasma was established as 99% deficient on average by ELISA testing. The albumin level serves as control for depletion process.
Catalog No. 50-274-3988 Supplier Innovative Research Inc Supplier No. ISAADPLY1ML
Add to Cart
Add to Cart
Serum Amyloid A (SAA) Deficient Human Plasma Lyophilized 1mL - Serum Amyloid A (SAA) Deficient Human Plasma Lyophilized from Innovative Research is supplied as a vial of depleted human plasma in lyophilized form. The plasma was established as 99% deficient on average by ELISA testing. The albumin level serves as control for depletion process.