Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ SAA (Human) ELISA Kit

Catalog No. 89032264 Shop All Abnova Corporation Products
Change view
Click to view available options
Target Species:
Human
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Target Species
89-032-264 Human
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-032-264 Supplier Abnova™ Supplier No. KA4865
Only null left
Add to Cart
Add to Cart

Sandwich enzyme immunoassay for the quantitative measurement of human SAA

  • Designed for detection of human SAA in plasma, serum, and cell culture samples
  • Measures total SAA in less than 4 hours

Specifications

Calibration Range 0.125 to 2.00 ug/mL
Description SAA (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human SAA.
Conjugate Biotin
Detection Method Colorimetric
Product Type ELISA Kit
Sample Type Cell Culture, Plasma, Serum
Target Species Human
For Use With (Application) Quantification
Gene ID (Entrez) 6288
Kit Contents Human Serum Amyloid A Microplate, Sealing Tapes, Human Serum Amyloid A Standard, Biotinylated Human Serum Amyloid A Antibody (50x), MIX Diluent Concentrate (10x), Wash Buffer Concentrate (20x), Streptavidin-Peroxidase Conjugate (SP Conjugate), Chromogen Substrate, Stop Solution: A 0.5 N hydrochloric acid
Regulatory Status For research use only (RUO)
Sample Volume 50 μL
Storage Requirements Store components of the kit at 4°C or -20°C as described in the protocol.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.