Learn More
Genscript Corporation SAA MLB2 mAb Mouse
Catalog No. 5000058448
Encompass
Serum Amyloid A (SAA) is an acute-phase protein The concentration of SAA in blood increases rapidly when tissue damage or inflammation occurs It is considered as an early biomarker for the diagnosis of many inflammatory diseases SAA can also be used as an important indicator of infection
Catalog No. 50-000-58448 Supplier Genscript Corporation Supplier No. V02006100
Add to Cart
Add to Cart
Request a Quote
Serum Amyloid A (SAA) is an acute-phase protein The concentration of SAA in blood increases rapidly when tissue damage or inflammation occurs It is considered as an early biomarker for the diagnosis of many inflammatory diseases SAA can also be used as an important indicator of infection