Learn More
ABclonal Technology SAA1 Rabbit pAb
SDP
Catalog No. 5000105017
Encompass_Preferred
This gene encodes a member of the serum amyloid A family of apolipoproteins. The encoded preproprotein is proteolytically processed to generate the mature protein. This protein is a major acute phase protein that is highly expressed in response to inflammation and tissue injury. This protein also plays an important role in HDL metabolism and cholesterol homeostasis. High levels of this protein are associated with chronic inflammatory diseases including atherosclerosis, rheumatoid arthritis, Alzheimers disease and Crohns disease. This protein may also be a potential biomarker for certain tumors. Finally, antimicrobial activity against S. aureus and E. coli resides in the N-terminal portion of the mature protein. Alternate splicing results in multiple transcript variants that encode the same protein. A pseudogene of this gene is found on chromosome 11.
Catalog No. 50-001-05017 Supplier ABclonal Technology Supplier No. A145531000UL
This gene encodes a member of the serum amyloid A family of apolipoproteins. The encoded preproprotein is proteolytically processed to generate the mature protein. This protein is a major acute phase protein that is highly expressed in response to inflammation and tissue injury. This protein also plays an important role in HDL metabolism and cholesterol homeostasis. High levels of this protein are associated with chronic inflammatory diseases including atherosclerosis, rheumatoid arthritis, Alzheimers disease and Crohns disease. This protein may also be a potential biomarker for certain tumors. Finally, antimicrobial activity against S. aureus and E. coli resides in the N-terminal portion of the mature protein. Alternate splicing results in multiple transcript variants that encode the same protein. A pseudogene of this gene is found on chromosome 11.