Learn More
Aviva Systems Biology Corp SAA4 Antibody - middle region (ARP42301_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42301_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SAA4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SAA4. Alternate names: C-SAA, CSAAApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Pig
Catalog No. 50-014-58176