Learn More
Aviva Systems Biology Corp SAAL1 Antibody - middle region (ARP52597_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52597_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SAAL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SAAL1. Alternate names: FLJ41463, SPACIA1ApplicationWB. Species reactivity: Guinea Pig, Horse, Human, Pig, Rat
Catalog No. 50-285-737