Learn More
Cayman Chemical SAAP 148trIfluoROCTAte sa 10mg
SDP
Catalog No. 502915855
Encompass_Preferred
An antimicrobial peptide; active against several multidrug-resistant bacteria in vitro; decreases the number of skin CFUs in mouse models of A. baumannii or MRSA infections at 0.125-2% w/w
Catalog No. 50-291-5855 Supplier Cayman Chemical Supplier No. 3751710 MG
An antimicrobial peptide; active against several multidrug-resistant bacteria in vitro; decreases the number of skin CFUs in mouse models of A. baumannii or MRSA infections at 0.125-2% w/w