Learn More
MOLTOX Molecular Toxicology Inc Sabouraud Dextrose Broth (SDB), pH 5.6 +/- 0.2, 100 mL/bottle, Store in Refrigerator
SDP
Catalog No. NC0803824
Encompass
This item has a minimum order quantity of 96 per supplier requirements
Sabouraud Dextrose Broth is used for culturing yeasts, molds and other aciduric microorganisms. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. NC0803824 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 26728
This item has a minimum order quantity of 96 per supplier requirements
Sabouraud Dextrose Broth is used for culturing yeasts, molds and other aciduric microorganisms. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.