Learn More
MilliporeSigma Sabouraud Dextrose Agar with neutralizers, 200/pk
Catalog No. NC1633673
Encompass
Sabouraud Dextrose Agar (SDA) is a complex medium for cultivation and isolation of yeasts and molds. The media finds its application in environmental monitoring by determining the yeast and mold count on surfaces in isolators and clean rooms.
Catalog No. NC1633673 Supplier MilliporeSigma Supplier No. 1465010200
Sabouraud Dextrose Agar (SDA) is a complex medium for cultivation and isolation of yeasts and molds. The media finds its application in environmental monitoring by determining the yeast and mold count on surfaces in isolators and clean rooms.