Learn More
Medchemexpress LLC Sabcomeline hydrochl 100mg | 159912-58-0 | 100MG
SDP
Catalog No. 5000242087
Encompass_Preferred
Sabcomeline (SB-202026) hydrochloride is a potent and functionally selective muscarinic M1 receptor partial agonist that improve cognition Sabcomeline hydrochloride can be used for Alzheimer s disease research[1 [2
Catalog No. 50-002-42087 Supplier Medchemexpress LLC Supplier No. HY106432A100MG
Sabcomeline (SB-202026) hydrochloride is a potent and functionally selective muscarinic M1 receptor partial agonist that improve cognition Sabcomeline hydrochloride can be used for Alzheimer s disease research[1 [2