Learn More
Medchemexpress LLC HY-108943 100mg Medchemexpress, Sabinene CAS:3387-41-5 Purity:>98%
SDP
Catalog No. 501968639
Encompass_Preferred
Medchemexpress, HY-108943 100mg Sabinene CAS:3387-41-5 Sabinene is a perfume additive which is being explored as the component for the next generation aircraft fuel. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8639 Supplier Medchemexpress LLC Supplier No. HY108943100MG
Medchemexpress, HY-108943 100mg Sabinene CAS:3387-41-5 Sabinene is a perfume additive which is being explored as the component for the next generation aircraft fuel. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.