Learn More
Medchemexpress LLC HY-108943 500mg Medchemexpress, Sabinene CAS:3387-41-5 Purity:98%
SDP
Catalog No. 501637415
Encompass_Preferred
Medchemexpress,HY-108943 500mg Sabinene CAS:3387-41-5 Formula:C10H16 Sabinene is a perfume additive which is being explored as the component for the next generation aircraft fuel. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-7415 Supplier Medchemexpress LLC Supplier No. HY108943500MG
Medchemexpress,HY-108943 500mg Sabinene CAS:3387-41-5 Formula:C10H16 Sabinene is a perfume additive which is being explored as the component for the next generation aircraft fuel. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.