Learn More
Medchemexpress LLC Sabizabulin | 1332881-26-1 | 99.0% | 100 MG
SDP
Catalog No. 5000234515
Encompass_Preferred
Sabizabulin (VERU-111) is an orally bioavailable small-molecule tubulin inhibitor that binds the colchicine site and demonstrates potent antiproliferative activity in preclinical studies, with reported antiviral and anti-inflammatory effects.
  • Oral tubulin inhibitor targeting the colchicine-binding site.
  • High purity (99.0%) and defined molecular weight (377.39 g·mol⁻¹).
  • Suitable for in vitro and in vivo research on microtubule dynamics and anticancer mechanisms.
  • Available in multiple sizes, including 100 mg, as a solid powder for research use.
  • For research use only; not for human therapeutic use.
Catalog No. 50-002-34515 Supplier Medchemexpress LLC Supplier No. HY120599100MG
Sabizabulin (VERU-111) is an orally bioavailable small-molecule tubulin inhibitor that binds the colchicine site and demonstrates potent antiproliferative activity in preclinical studies, with reported antiviral and anti-inflammatory effects.
  • Oral tubulin inhibitor targeting the colchicine-binding site.
  • High purity (99.0%) and defined molecular weight (377.39 g·mol⁻¹).
  • Suitable for in vitro and in vivo research on microtubule dynamics and anticancer mechanisms.
  • Available in multiple sizes, including 100 mg, as a solid powder for research use.
  • For research use only; not for human therapeutic use.