Learn More
Medchemexpress LLC Sabizabulin | 1332881-26-1 | 99.0% | 50 MG
SDP
Catalog No. 5000231936
Encompass_Preferred
Sabizabulin (VERU-111; ABI-231) is a small-molecule tubulin inhibitor provided for research use. It disrupts microtubule dynamics and is commonly used in cellular assays and mechanism-of-action studies. Reported purity is 99.0% and the compound is available in multiple small-mass packaging options.
  • Small-molecule tubulin inhibitor used in cellular and biochemical assays.
  • Reported purity 99.0% for consistent experimental results.
  • Available in small-mass packaging suitable for preclinical studies.
  • Useful for mechanism-of-action and microtubule dynamics research.
  • CAS number 1332881-26-1 for unambiguous identification.
Catalog No. 50-002-31936 Supplier Medchemexpress LLC Supplier No. HY12059950MG
Sabizabulin (VERU-111; ABI-231) is a small-molecule tubulin inhibitor provided for research use. It disrupts microtubule dynamics and is commonly used in cellular assays and mechanism-of-action studies. Reported purity is 99.0% and the compound is available in multiple small-mass packaging options.
  • Small-molecule tubulin inhibitor used in cellular and biochemical assays.
  • Reported purity 99.0% for consistent experimental results.
  • Available in small-mass packaging suitable for preclinical studies.
  • Useful for mechanism-of-action and microtubule dynamics research.
  • CAS number 1332881-26-1 for unambiguous identification.