Learn More
Ted Pella Inc Sable Brushes, #000, 1.0mm W x 6.0mm L, 3/pk
SDP
Catalog No. NC0258565
Encompass
For precise manipulation of TEM specimen grids and samples, dusting of SEM samples and photographic materials or photographic spotting. Sable hairs are springy and relatively fine. Length of hairs are short for better manipulation. Enameled wood handles and seamless nickel ferrules.
Catalog No. NC0258565 Supplier Ted Pella Inc Supplier No. 11807
For precise manipulation of TEM specimen grids and samples, dusting of SEM samples and photographic materials or photographic spotting. Sable hairs are springy and relatively fine. Length of hairs are short for better manipulation. Enameled wood handles and seamless nickel ferrules.