Learn More
Ted Pella Inc Sable Brushes, #00000, 0.08mm W x 4.0mm L, 3/pk
SDP
Catalog No. NC9052674
Encompass
For precise manipulation of TEM specimen grids and samples, dusting of SEM samples and photographic materials or photographic spotting. Sable hairs are springy and relatively fine. Length of hairs are short for better manipulation. Enameled wood handles and seamless nickel ferrules. Offered in packages of 3 brushes. Measurements taken just above ferrule.
Catalog No. NC9052674 Supplier Ted Pella Inc Supplier No. 11806
For precise manipulation of TEM specimen grids and samples, dusting of SEM samples and photographic materials or photographic spotting. Sable hairs are springy and relatively fine. Length of hairs are short for better manipulation. Enameled wood handles and seamless nickel ferrules. Offered in packages of 3 brushes. Measurements taken just above ferrule.