Learn More
Ted Pella Inc SABLE BRUSHES NO1 PKG/3
SDP
Catalog No. NC9361368
Encompass
Brushes; Sable; Red; Width at Ferrule 1.5mm, Length at ferrule 9.5mm, no.1; Sable hairs are springy and relatively fine, which gives them a special place in the microscopist's tools armory; 3/PK
Catalog No. NC9361368 Supplier Ted Pella Inc Supplier No. 11812
Brushes; Sable; Red; Width at Ferrule 1.5mm, Length at ferrule 9.5mm, no.1; Sable hairs are springy and relatively fine, which gives them a special place in the microscopist's tools armory; 3/PK