Learn More
NEOGEN Sabouraud Dextrose Agar, Emmons, 10 kg
SDP
Catalog No. 502015908
Encompass_Preferred
Sabouraud Dextrose Agar, Emmons is used for the cultivation of fungi in a laboratory setting. Sabouraud Dextrose Agar, Emmons is not intended for use in the diagnosis of disease or other conditions in humans. Sabouraud Dextrose Agar (SDA), Emmons is a modification of Dextrose Agar described by Sabouraud. SDA is used for cultivating pathogenic and commensal fungi and yeasts. Emmons modified the formula by adjusting the pH and reducing the dextrose concentration. This neutral pH enhances recovery of some pathogenic fungi. Sabouraud Dextrose Agar, Emmons is an excellent basal medium, and antibiotics and inhibitors may be added for the selective cultivation and isolation of various microorganisms.For more information visit https://www.neogen.com/solutions/Microbiology/sabouraud-dextrose-agar-emmons
Catalog No. 50-201-5908 Supplier NEOGEN Supplier No. 700003622
Sabouraud Dextrose Agar, Emmons is used for the cultivation of fungi in a laboratory setting. Sabouraud Dextrose Agar, Emmons is not intended for use in the diagnosis of disease or other conditions in humans. Sabouraud Dextrose Agar (SDA), Emmons is a modification of Dextrose Agar described by Sabouraud. SDA is used for cultivating pathogenic and commensal fungi and yeasts. Emmons modified the formula by adjusting the pH and reducing the dextrose concentration. This neutral pH enhances recovery of some pathogenic fungi. Sabouraud Dextrose Agar, Emmons is an excellent basal medium, and antibiotics and inhibitors may be added for the selective cultivation and isolation of various microorganisms.For more information visit https://www.neogen.com/solutions/Microbiology/sabouraud-dextrose-agar-emmons