Learn More
MilliporeSigma Sabouraud Dextrose Agar, 120/pk
Catalog No. NC0740187
Encompass
Sabouraud Dextrose Agar settle plate is designed for the detection of the total count of yeasts and molds in air and from personnel in less critical environments.
Catalog No. NC0740187 Supplier MilliporeSigma Supplier No. 1460280120
Sabouraud Dextrose Agar settle plate is designed for the detection of the total count of yeasts and molds in air and from personnel in less critical environments.