Learn More
Research Products International Corp Sabouraud Dextrose Agar, Kilogram
SDP
Catalog No. 50488992
Encompass_Preferred
Sabouraud Dextrose Agar is an agar medium recommended for the isolation, cultivation and identification of non-pathogenic and pathogenic fungi. This medium does not contain antibiotics or inhibitors and can be used for the cultivation of those fungi which are susceptible to antibiotics.
Dissolve 65.0 grams in 1 liter distilled water. Sterilize by autoclaving for 15 minutes.
Ingredients:
Casein Digest Peptone: 10.0 g/L
Dextrose: 40.0 g/L
Agar: 15.0 g/L
Catalog No. 50-488-992 Supplier Research Products International Corp Supplier No. S125001000.0
Sabouraud Dextrose Agar is an agar medium recommended for the isolation, cultivation and identification of non-pathogenic and pathogenic fungi. This medium does not contain antibiotics or inhibitors and can be used for the cultivation of those fungi which are susceptible to antibiotics.
Dissolve 65.0 grams in 1 liter distilled water. Sterilize by autoclaving for 15 minutes.
Ingredients:
Casein Digest Peptone: 10.0 g/L
Dextrose: 40.0 g/L
Agar: 15.0 g/L