Learn More
Alkali Scientific Sabouraud Dextrose Agar, 2 Kg
SDP
Catalog No. 5000660236
Encompass_Preferred
Sabouraud Dextrose Agar (SDA) is a widely used fungal growth medium that supports the cultivation of yeasts, molds, and other fungi. This 2 kg package is ideal for large-scale testing and environmental monitoring, providing a high-volume supply for laboratories focused on isolating fungi from clinical and environmental samples. The medium contains dextrose as a primary energy source for fungi and maintains an acidic pH to inhibit bacterial growth, making it ideal for fungal isolation. SDA is used in clinical diagnostics, cosmetic testing, food industry applications, and environmental monitoring, where fungal contamination must be assessed and controlled.
Catalog No. 50-006-60236 Supplier Alkali Scientific Supplier No. BOU002CP
Sabouraud Dextrose Agar (SDA) is a widely used fungal growth medium that supports the cultivation of yeasts, molds, and other fungi. This 2 kg package is ideal for large-scale testing and environmental monitoring, providing a high-volume supply for laboratories focused on isolating fungi from clinical and environmental samples. The medium contains dextrose as a primary energy source for fungi and maintains an acidic pH to inhibit bacterial growth, making it ideal for fungal isolation. SDA is used in clinical diagnostics, cosmetic testing, food industry applications, and environmental monitoring, where fungal contamination must be assessed and controlled.