Learn More
Alkali Scientific Sabouraud Dextrose Agar, 20 mL Deep Pour, 20 × 125 mm Tube, 10/Pk
SDP
Catalog No. 5000662860
Encompass_Preferred
Sabouraud Dextrose Agar, 20 mL Deep Pour, 20 × 125 mm Tube, 10/Pk. Ideal for cultivating fungi, this agar provides a deep pour design for enhanced sample analysis. The 20 mL tube ensures easy application and is perfect for microbiological research. The product comes in a convenient pack of 10, ensuring plenty of supply for long-term use.
Catalog No. 50-006-62860 Supplier Alkali Scientific Supplier No. SDADT00
Sabouraud Dextrose Agar, 20 mL Deep Pour, 20 × 125 mm Tube, 10/Pk. Ideal for cultivating fungi, this agar provides a deep pour design for enhanced sample analysis. The 20 mL tube ensures easy application and is perfect for microbiological research. The product comes in a convenient pack of 10, ensuring plenty of supply for long-term use.