Learn More
MOLTOX Molecular Toxicology Inc Sabouraud Dextrose Agar, 20 mL/plate, 20 plates/sleeve, Store at Room Temperature
SDP
Catalog No. 50948672
Encompass_Preferred
Sabouraud Dextrose Agar is used for the cultivation of pathogenic and nonpathogenic fungi such as dermatophytes and can be more selective with the addition of antimicrobics. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-672 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 2140S53
Sabouraud Dextrose Agar is used for the cultivation of pathogenic and nonpathogenic fungi such as dermatophytes and can be more selective with the addition of antimicrobics. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.