Learn More
Alkali Scientific Sabouraud Dextrose Agar, 500 G
SDP
Catalog No. 5000660782
Encompass_Preferred
Sabouraud Dextrose Agar (SDA) is the standard medium for cultivating and isolating yeasts and molds. This 500g package is ideal for laboratories that require medium-scale fungal culture and isolation for clinical diagnostics, environmental testing, and quality control in the food industry. The formulation provides a high concentration of dextrose, supporting robust fungal growth, and the slightly acidic pH (around 5.6) discourages bacterial growth, ensuring selective fungal isolation. The medium is widely used for isolating pathogens, environmental monitoring, and maintaining stock fungal cultures, especially in pharmaceutical and clinical microbiology applications.
Catalog No. 50-006-60782 Supplier Alkali Scientific Supplier No. BOU003CP
Sabouraud Dextrose Agar (SDA) is the standard medium for cultivating and isolating yeasts and molds. This 500g package is ideal for laboratories that require medium-scale fungal culture and isolation for clinical diagnostics, environmental testing, and quality control in the food industry. The formulation provides a high concentration of dextrose, supporting robust fungal growth, and the slightly acidic pH (around 5.6) discourages bacterial growth, ensuring selective fungal isolation. The medium is widely used for isolating pathogens, environmental monitoring, and maintaining stock fungal cultures, especially in pharmaceutical and clinical microbiology applications.