Learn More
Teknova Sabouraud Dextrose Agar. 500mL, Sterile. 4 pack.
SDP
Catalog No. 502387291
Encompass
1.0% Tryptone, 4.0% Cerelose Anhydrous Dextrose (Glucose), 1.5% Agar
Catalog No. 50-238-7291 Supplier Teknova Supplier No. S87014PK
1.0% Tryptone, 4.0% Cerelose Anhydrous Dextrose (Glucose), 1.5% Agar