Learn More
NEOGEN Sabouraud Dextrose Agar, 5 kg
SDP
Catalog No. 502015175
Encompass_Preferred
Sabouraud Dextrose Agar is used for the cultivation of fungi. Conforms to Harmonized USP/EP/JP performance requirements and is not intended for use in the diagnosis of disease or other conditions in humans. A medium recommended by the Harmonized Pharmacopeia for the isolation and identification of Candida albicans from non-sterile samples. Conforms to USP/EP/JP performance specification. The medium is commonly abbreviated to SDA. The medium is also used for the preparation and maintenance of fungal test strains as described by the Harmonized Pharmacopeia. The peptone digests and dextrose provide a nutritious base for luxuriant fungal growth and the acidic pH affords selectivity against bacteria. Due to the high carbohydrate content and low pH this medium is highly sensitive to overheating which will cause a drop in the gel strength.For more information visit https://www.neogen.com/solutions/Microbiology/sabouraud-dextrose-agar
Catalog No. 50-201-5175 Supplier NEOGEN Supplier No. 700002980
Sabouraud Dextrose Agar is used for the cultivation of fungi. Conforms to Harmonized USP/EP/JP performance requirements and is not intended for use in the diagnosis of disease or other conditions in humans. A medium recommended by the Harmonized Pharmacopeia for the isolation and identification of Candida albicans from non-sterile samples. Conforms to USP/EP/JP performance specification. The medium is commonly abbreviated to SDA. The medium is also used for the preparation and maintenance of fungal test strains as described by the Harmonized Pharmacopeia. The peptone digests and dextrose provide a nutritious base for luxuriant fungal growth and the acidic pH affords selectivity against bacteria. Due to the high carbohydrate content and low pH this medium is highly sensitive to overheating which will cause a drop in the gel strength.For more information visit https://www.neogen.com/solutions/Microbiology/sabouraud-dextrose-agar