Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ Sabouraud Dextrose Agar with Chloramphenicol
SDP

Catalog No. 23001769 Shop All Hardy Diagnostics Products
Change view
Click to view available options
No. per Pack:
10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
23-001-769 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-769 Supplier Hardy Diagnostics™ Supplier No. W72
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sabouraud Dextrose Agar with Chloramphenicol is recommended for the selective isolation of fungi and yeasts from clinical and nonclinical specimens

  • Standard size (15X100mm) passive settling plate for air monitoring of bacteria or fungi
  • Ideal for environmental monitoring in a clean room or clean bench setting
  • Optically clear polystyrene for distortion free viewing
  • Polystyrene Petri plates feature raised rings for stacking ability and prevents sliding

Specifications

Appearance Translucent
Color Light Amber
Additive Chloramphenicol
Detects Fungi, Yeast, Mold
Form Prepared Medium
Format Agar Plate
Media Type Sabouraud Dextrose Agar
Product Type Agar Plate
Target Organism Class Fungi
Test Type Fungal Testing
Fill Volume 26 mL
For Use With (Application) Selective isolation of fungi and yeasts from clinical and nonclinical specimens, Cannabis QA Testing, Clinical, Environmental, Food Safety, Pharmaceutical Manufacturing
Incubation Conditions 20°C to 25°C, Aerobic
Incubation Time Variable
Material Polystyrene
No. per Pack 10
Packaging Type Sleeves, Breathable Bag
Quantity 10/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.