Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ Sabouraud Dextrose Agar with Lecithin and Tween™ 80
SDP

Catalog No. 23001785 Shop All Hardy Diagnostics Products
Change view
Click to view available options
No. per Pack:
10 Bottles
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
23-001-785 10 Bottles
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-785 Supplier Hardy Diagnostics™ Supplier No. U175
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sabouraud Dextrose Agar with Lecithin and Tween 80 is recommended for the cultivation of yeasts and fungi from environmental samples

  • Sabouraud Dextrose Medium contains digests of animal tissues (peptones) which provide a nutritious source of amino acids and nitrogenous compounds for the growth of fungi and yeasts
  • Dextrose is added as the energy and carbon source
  • Lecithin and Tween™ 80 are added to neutralize the antimicrobial effects of disinfectants or cleaning solutions used on environmental surfaces
  • The pH is adjusted to approximately 5.6 in order to enhance the growth of fungi, especially dermatophytes

Specifications

Appearance Translucent
Color Light Amber
Additive Lecithin, Tween 80
Detects Fungi
Form Prepared Medium
Format Bottle
Media Type Agar
Preparation Method Ready-to-Use
Product Type Agar
Target Organism Class Yeast
Test Type Microbial Cultivation
Fill Volume 400 mL
Final Product Type Microorganisms
For Use With (Application) Cultivation of Fungi from Environmental Samples
Material Polycarbonate
No. per Pack 10 Bottles
Packaging Type Bottle
Specimen Type Environmental Samples
Volume (Metric) Specimen 400 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.