Learn More
Teknova Sabouraud Dextrose Agar Plates. 150mm Plates, Sterile. 4 pack.
SDP
Catalog No. 502387401
Encompass
Sabouraud dextrose agar is a selective medium used to isolate yeast and fungi while permitting the growth of filamentous bacteria like Nocardia.
Catalog No. 50-238-7401 Supplier Teknova Supplier No. S20504PK
Sabouraud dextrose agar is a selective medium used to isolate yeast and fungi while permitting the growth of filamentous bacteria like Nocardia.