Learn More
Teknova Sabouraud Dextrose Agar Plates. 100mm, 20 Plates per Sleeve, Sterile.
SDP
Catalog No. 502387968
Encompass
Sabouraud dextrose agar is a selective medium used to isolate yeast and fungi while permitting the growth of filamentous bacteria like Nocardia.
Catalog No. 50-238-7968 Supplier Teknova Supplier No. S2040
Sabouraud dextrose agar is a selective medium used to isolate yeast and fungi while permitting the growth of filamentous bacteria like Nocardia.